KIMBLE® KONTES® Hybridization Tubes, DWK Life Sciences
Proveedor: DWK Life Sciences
KONTES®
736500-3530 736500-3515
KT736500-3515PK 4999.34 MXN
KT736500-3515 KT736500-3530
KIMBLE® KONTES® Hybridization Tubes, DWK Life Sciences
Frascos Botellas para hibridación
For use with hybridization incubators with rotators.
  • Tubes significantly reduce probe volumes
Perform hybridization of multiple Northern, Southern, or Western blots (up to five per tube) in a single tube, using nylon mesh sheets to separate the hybridization membranes.
Información para pedidos: Complete with 45 mm polypropylene caps and silicone O-rings.