KIMAX® Berzelius Beakers, Tall Form, Graduated, Borosilicate Glass, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
14030-100 14020-200 14030-200 14020-400 14020-100 14020-1000 14030-300 14030-400 14020-300 14020-600 14030-1000 14030-600
89000-748CS 8747.14 MXN
89000-748 89001-088 89001-086 13920-137 89001-090 89000-750 13920-034 13920-078 13920-012 13920-057 89000-752 89000-698
KIMAX® Berzelius Beakers, Tall Form, Graduated, Borosilicate Glass, Kimble Chase
Vasos de precipitados
These berzelius, tall form, double capacity scale beakers are manufactured with uniform wall thickness, particularly in the critical radius point of the heel area.
  • Beakers feature a slightly flared top with a thick reinforcing bead
  • Available with or without a pouring spout
  • All sizes have a durable matte finish area for clear marking, and easy-to-read white scales to indicate approximate volumes
This strong and durable beaker exhibits excellent mechanical and thermal shock resistance.
Certificaciones: Beakers with a pouring spout are designed from ASTM Specification E960, Type III requirements; beakers without a pouring spout are designed from ASTM Specification E960, Type IV requirements.