Technik™ Stainless Steel Cutters, Aven Tools
Proveedor: Aven
Technik™
10321 10323
47743-092EA 805.9 MXN
47743-092 47743-094
Technik™ Stainless Steel Cutters, Aven Tools
Cortadores
These strong and versatile oval head cutters are extremely popular and used for general purpose wire cutting.
  • Box joint construction provides superior jaw alignment and durability
  • Induction hardened to 50-52 HRC
Dual leaf springs ensure opening and closing is comfortable. ESD Safe Grips provides protection against static as well as extra comfort for repetitive work. Mirror finished stainless steel (400 series) provides superior rust protection compared with standard carbon steel pliers.