Corning® Polystyrene Tissue Culture Flasks, Sterile, Corning
Proveedor: Corning
Corning®
430168 431079 430372 430823 431081 431085 430824
29186-024CS 16500 MXN
29186-002 89090-948 29186-024 29186-199 29186-160 29186-192 29560-969
Corning® Polystyrene Tissue Culture Flasks, Sterile, Corning
Matraces Matraces de cultivo de células
Designed and treated for optimal cell attachment and growth
  • Certified nonpyrogenic
  • The lot number is printed on all flasks for traceability
  • Canted models have a skirt to prevent tipping
Flasks 29442-028 and 29442-020 are triangular in shape for increased stability and pipet access. All other flasks are either standard rectangular or U-shaped.
The original plug-seal polyethylene cap can be used in both open and closed systems. Phenolic-style polyethylene caps are designed for open gas exchange applications.