Corning® Polystyrene Flasks with Vented Cap, Sterile, Corning
Proveedor: Corning
Corning®
3151
29442-004CS 3049.7 MXN
29442-004
Corning® Polystyrene Flasks with Vented Cap, Sterile, Corning
Matraces Matraces de cultivo de células
Treated for optimum cell attachment and growth
Polyethylene cap contains a 0.2µm hydrophobic filter to allow gas exchange and minimize contamination. Certified nonpyrogenic. Lot number is printed on flasks for traceability.