Corning® Polystyrene Tissue Culture Flasks with Vented Caps, Sterile, Corning
Proveedor: Corning
Corning®
430825 431082 430639 431080 431306
29560-959CS 5400 MXN
29560-959 29186-010 10010-366 89090-950 29186-106
Corning® Polystyrene Tissue Culture Flasks with Vented Caps, Sterile, Corning
Matraces Matraces de cultivo de células
These flasks are tissue culture-treated for optimum cell attachment and growth
  • Certified nonpyrogenic
  • Lot number is printed on each flask for traceability
  • Canted flasks have a skirt to prevent tipping
The triangular flasks have the shape for increased stability and pipet access. The flasks packaged in cases of 84 come prelabeled with a bar code and is validated for use with Select Robotic System.
Polyethylene caps contain a 0.2 µm hydrophobic filter to allow gas exchange and minimize risk of contamination.