TouchNTuff® 92-600, Nitrile Gloves, Powder-Free, Ansell
Proveedor: Ansell
TouchNTuff®
105077 585835 9260000M-VEN 9260000L-VEN 9260011080 9260011090 9260011100 9260000XL-VEN 585836 585834 585837 9260000S-VEN
34800-500CS 4473 MXN
34800-500 CA76006-390 CA34800-506 CA76006-396 CA34800-504 34800-506 CA76006-392 CA34800-502 34800-504 CA34800-500 34800-502 CA76006-394
TouchNTuff® 92-600, Nitrile Gloves, Powder-Free, Ansell
Guantes
The TouchNTuff 92-600 gloves are thin (4 mil), 100% nitrile disposable gloves which are applicable for chemical splash protection.
  • Superior puncture resistance
  • Exceptional durability and fingertip sensitivity
  • Silicone-free
They offer three times the puncture resistance of standard latex or vinyl gloves. These gloves combine excellent fingertip sensitivity with exceptional durability to give a high level of protection against grease and oil. They are lightweight and comfortable to wear.
Certificaciones: Touch N Tuff® gloves comply with FDA food handling requirements (21 CFR 177.2600).