Nalgene® Square Media Bottles, PETG, Sterile, Graduated, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
2019-2000 2019-0030 2019-0500 2019-0125 2019-1000 2019-0060 2019-0250
16159-132CS 6442.13 MXN
16159-132 16159-130 16159-136 16159-134 16159-128 16159-138 16159-126
Nalgene® Square Media Bottles, PETG, Sterile, Graduated, Thermo Scientific
Frascos Frascos de medios
Graduated polyethylene terephthalate copolyester bottles are inexpensive alternatives to glass media bottles.
  • They are noncytotoxic
  • Comply with USP Class VI guidelines
Media bottles are radiation sterilized (SAL of 10⁻⁶) and nonpyrogenic to eliminate costly washing, depyrogenation, and autoclaving steps. A heat-shrink band around the closure and neck provide a tamper-evident seal.
These heavy-walled, break-resistant, durable bottles are transparent and offer reduced permeability to CO₂ and O₂.
Información para pedidos: Packed in shrink wrapped trays and sold in case quantities only. The 2 L PETG bottle is equipped with molded-in handgrips and a 53 mm (53B) white closure.