VWR® Erlenmeyer Flasks, Narrow Mouth, Reinforced Beaded Top, Capacity Scale
Proveedor: Avantor
VWR®
VW26500-1000
89000-368CS 7082.31 MXN
89000-368
VWR® Erlenmeyer Flasks, Narrow Mouth, Reinforced Beaded Top, Capacity Scale
Matraces Matraces Erlenmeyer
Flasks are versatile and economic, with a thick-walled body and long, tapered outside contour to minimize chipping.
  • Ideal for general laboratory use
  • Thick-walled body
  • Versatile and economic option
Tops are tooled with a rounded finish and reinforced for maximum mechanical strength.
Meet ASTM Specification E1404, Type I, Class 1 requirements.
Durable white ceramic enamel scales indicate approximate volumes for measuring and mixing.