Nalgene® Square Media Bottles, PET, Sterile, Graduated, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
342040-0125 342044-0650 342040-1000 342044-1000 342040-0250 342040-0650 342044-0125
16010-100CS 3396.1 MXN
16010-100 73521-216 16010-104 73520-444 73520-442 16010-102 16010-106
Nalgene® Square Media Bottles, PET, Sterile, Graduated, Thermo Scientific
Frascos Frascos de medios
Bottles are lightweight, shatter-resistant, and provide excellent gas barrier properties for storage and shipping of liquid media, buffers, and sera.
  • Noncytotoxic
Sterile and nonpyrogenic to save time and prevent costly cleaning preparation. Bottles feature molded-in graduations and molded-in neck ring for use with tamper-evident shrink sleeves. Suitable for air shipment and tested to ensure leakproof performance.
Certificaciones: Bottles meet USP Class VI, USP 661, and European Pharmacopeia for Abnormal Toxicity standards.
Información para pedidos: Packaged in shrink-wrapped trays. Trays are double-bagged for critical environment use. Bottles are packed 24 per tray, except bottles 16010-104 and 16010-106, which are packed 12 per tray. All bottles feature a 38-430 neck finish; a high-density polyethylene closure is included where indicated.