VWR® Boston Round Media Bottles, PC
Proveedor: VWR
VWR®
RPC0300 RPC1000 RPC0500
89047-227CS 27267.98 MXN
30180-028 89047-227 30180-030
VWR® Boston Round Media Bottles, PC
Frascos Frascos de medios
VWR® polycarbonate resin is an amorphous engineering thermoplastic, characterized by outstanding mechanical, optical, electrical, and thermal properties.
  • Polycarbonate resin construction
  • Autoclavable
  • Optimal for preparation of terminally sterile fluids
The Lexan® resins employed are environmentally conforming to flame retardancy, scratch resistance, toughness, heat resistance, weatherability, biocompatibility, optical quality, and compliance with stringent FDA and USP requirements.
All bottles are manufactured and packaged in a Class 10K cleanroom in an ISO 9001 compliant facility, and do not require rinsing prior to first use.
A safe alternative to glass vessels, these media bottles are ideal for steam sterilization of all biologicals, liquids, reagents, media, buffers, and aqueous solutions. Media bottles withstand repeated autoclaving, and are optimal for all microbial culture media production and preparation of terminally sterile fluids.
Información para pedidos: Bottles are supplied nonsterile with standard blue autoclavable caps attached. Packaged in multi-barrier elastomer packaging. Caps are available with custom color options on bulk orders. For additional cap selections, see 82013-158 series.