CR100® Sterile Nitrile Gloves, Cardinal
Proveedor: Cardinal
CR100®
2Y1866T 2Y1864T 2Y1864T 2Y1862T 2Y1863T 2Y1861T 2Y1867T 2Y1863T 2Y1865T 2Y1866T 2Y1860T 2Y1867T 2Y1865T
PH2Y1862TCS 7481.88 MXN
PH2Y1862T CAPH2Y1863T CAPH2Y1864T PH2Y1861T PH2Y1864T CAPH2Y1865T CAPH2Y1866T PH2Y1863T CAPH2Y1860T PH2Y1866T CAPH2Y1867T PH2Y1865T PH2Y1867T
CR100® Sterile Nitrile Gloves, Cardinal
Guantes
Sterile cleanroom gloves are designed for use in biotechnology or pharmaceutical applications.
  • Beaded cuff resists roll-down for easy glove donning
  • Hand-specific
  • Powder-free
Strong, puncture-resistant nitrile formulation contains no natural rubber latex. Microtexture finish provides a sure grip. Color: white.
Certificaciones: Gamma irradiated to 10-6 sterility assurance level. Meet or exceed ASTM D6319 requirements for tensile strength.
Información para pedidos: Clean-processed and clean-packed in an ISO Class 4 (FED-STD-209E Class 10/M2.5) environment. Pairs are individually packed in a polyfilm inner wallet inside an easy-tear outer poly bag. Lot number is embossed on the poly bag edge for full traceability.