Falcon® Cell Culture Flasks, Corning
Proveedor: Corning
Falcon®
353018 353112 353107 353136 353108 355000 353139 353014 353112 353139 353082 353107 355000 353109 353009 353133 353109 353133 353110 353009 353135 355001 353028 355001 353138 353024 353082 353135 353110 353014 353028 353138 353018 353024 353108 353136
29185-060CS 3885.98 MXN
15705-072 CABD353082 29185-060 29185-085 29185-087 CA29185-298 29185-300 CA29185-078 29185-302 15705-074 29185-304 CA29185-258 29185-308 CABD353133 BD353136 CABD353135 BD353135 CABD353136 BD353133 CABD353138 CABD353139 47743-882 47743-884 CA29185-060 CA29185-085 29185-053 29185-298 29185-078 CA29185-087 CA15705-074 29185-258 CA29185-300 CA15705-072 CA29185-302 CA29185-304 CA29185-308
Falcon® Cell Culture Flasks, Corning
Matraces Matraces de cultivo de células
PS, tissue culture-treated, sterile, non-pyrogenic.
  • Easy to read printed volumetric graduations, and writing patch
  • Vented caps incorporate a 0.2 µm hydrophobic membrane
  • Convenient, peel-open, medical-style packaging ensures flask sterility is maintained
Vacuum gas plasma tissue culture treatment ensures consistent cell attachment, spreading and growth.
Barcoded (robotics/automation compatible).
Precaución: NOTE: Culture area and capacity are nominal.