PYREX® Distilling Flasks, Angle Type, 3 Necks, [ST] Joints, Corning
Proveedor: Corning
PYREX®
4965B-250 4965A-500 4965A-1L 4965-500 4965-1L 4965-100 4965B-500
89091-430CS 14400 MXN
89091-080 89091-216 29281-131 89091-430 32645-390 32645-392 32645-391
PYREX® Distilling Flasks, Angle Type, 3 Necks, [ST] Joints, Corning
Matraces Matraces de destilación
PYREX® borosilicate glass, with angled side necks, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One center neck and two side necks. Three necks with two angled side necks at the same height with 24/40 standard taper outer joints and a larger center neck with a vertical 34/45 standard taper joint. Matt white labelling area.