PYREX® Wide Mouth Media Storage Bottles, Corning
Proveedor: Corning
PYREX®
1397-500 1397-2L 1397-1L
89090-062CS 7000 MXN
89090-062 89090-060 89090-054
PYREX® Wide Mouth Media Storage Bottles, Corning
Frascos Frascos de medios
Made of PYREX® borosilicate glass.
  • Resistant to thermal shock
  • Permanent white enamel graduations and marking spot
  • Traceability Code for easier documentation
  • Not recommended for vaccum use
Wide mouth media storage bottles are designed for heavy duty storage as well as mixing and sampling. They includes linerless, one-piece autoclavable orange polypropylene plug seal GLS80-threaded cap with a drip-free pouring ring.
Precaución: Do not tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. These bottles should not be used in applications involving vacuum pressure as breakage may occur.