B.O.D. Bottles, 300 ml, KIMBLE®, DWK Life Sciences
Proveedor: DWK Life Sciences
15070-00 15070-25 15070-01
16429-602CS 6839.2 MXN
16429-604 16429-602 16429-600
B.O.D. Bottles, 300 ml, KIMBLE®, DWK Life Sciences
Frascos Frascos de DBO
Bottles are designed for sampling and incubation of aqueous samples for B.O.D.
Bottle’s shoulder radius provides an interior shape which sweeps entrained air out of the stopper opening. Stopper design uses a conical extension to displace excess sample which further ensures no air entrapment in the sample.
analysis. For reference see Method 5210-B: 5-day B.O.D. Test, Standard Methods for Examination of Water and Wastewater. Bottles conform to ASTM E438, Type I, Class B, borosilicate glass.