KIMBLE® RAY-SORB® Amber Erlenmeyer Flask, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMBLE® RAY-SORB®
26610-500 26610-50 26610-125 26610-250
16143-160EA 1272.12 MXN
16143-160 89001-352 16143-154 16143-156
KIMBLE® RAY-SORB® Amber Erlenmeyer Flask, DWK Life Sciences
Matraces Matraces Erlenmeyer
These flasks are RAY-SORB® processed to protect solutions sensitive to light from shorter wavelengths.
No capacity scale provided. Designed from ASTM Specification E1404, Type II, Class 2 requirements.