PYREX® Filtering Flasks, Heavy Wall with Tubulation, Corning
Proveedor: Corning
PYREX®
5340-4L 5340-250 5340-500 5360-50 5340-2L 5360-125 5340-1L 5360-25
89090-778CS 12821.89 MXN
89090-920 89090-778 89090-856 89090-736 89090-868 89090-824 89090-576 89090-760
PYREX® Filtering Flasks, Heavy Wall with Tubulation, Corning
Matraces Matraces de filtro
PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks have a beaded top that provides an excellent fit for rubber stoppers. Graduated to show approximate volume.
The heavy-duty rim flange is ideal for supporting Buchner funnels. It includes a side arm for attaching rubber tubing. Extra-large marking spot for easy identifications.