PYREX® VISTA™ Micro Filtering Flasks, Corning
Proveedor: Corning
PYREX® VISTA™
70360-50 70360-125 70360-25
89090-314CS 9843.83 MXN
89090-314 89090-358 89090-396
PYREX® VISTA™ Micro Filtering Flasks, Corning
Matraces Matraces de filtro
Made of borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These small filtering flasks are recommended for microchemical use. All flasks have permanent blue enamel marking spots.
Tubulation outer Ø: 10 mm
Certificaciones: ASTM E-1406 Type 2