KIMAX® Kjeldahl Flasks, Kimble Chase
Proveedor: DWK Life Sciences
KIMAX®
27400-100 27400-800 27400-30 27400-500
89001-850CS 17813.43 MXN
89001-850 89001-848 89001-844 89001-842
KIMAX® Kjeldahl Flasks, Kimble Chase
Matraces Matraces Kjeldahl
All flasks have reinforcing beads at the top
The 500mL and 800mL flasks have tooled necks to provide for accurate fit of stoppers and to reduce possibility of breakage. The 30mL size is designed from ASTM Specification E147. The 100mL and larger sizes are designed from ASTM Specification E1377, Type I requirements.