VWR® Digital Hygrometers/Thermometers with Probe
Proveedor: VWR
VWR®
4080 90011-35 VC4080
35519-041EA 18489.24 MXN
10754-552 35519-041 35519-020
VWR® Digital Hygrometers/Thermometers with Probe
Higrómetros Termohigrómetros digitales
These portable instruments are ideal for cleanrooms, hoods, environmental chambers, corrosion analysis, food storage, drug manufacturing, process control, incubators, desiccators, and all inside/outside air monitoring.
  • Traceable®
  • For laboratory, plant, and field use
To ensure accuracy, a 1µm thick, dielectric-polymer layer absorbs water molecules through a thin-metal electrode and causes a capacitance change proportional to relative humidity
Large, bright, four-digit, LCD display. Easy to use — no wicks to wet, no warm-up, no swinging, just point and read. Rugged ABS (acrylonitrile-butadiene-styrene) exterior designed for years of precise, dependable service.
A thermistor senses temperature.
Certificaciones: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Información para pedidos: Complete with probe, 9V alkaline battery, low-battery indicator, ABS case, Traceable certificate, and carrying case.