VWR® Traceable® Hygrometer/Thermometer/Barometer/Dew Point Pen
Proveedor: VWR
VWR® Traceable®
VC4247 93676-05
89094-760EA 7072.66 MXN
89094-760 10754-858
VWR® Traceable® Hygrometer/Thermometer/Barometer/Dew Point Pen
Higrómetros Termohigrómetros digitales
Compact design features an electronic capacitance polymer film sensor that is not affected by condensation.
  • Traceable®
  • Temperature in °C and °F
  • Compact design
  • Tough ABS plastic housing
Tough ABS plastic housing and water-resistant design with O-ring seal and membrane keypad assure a long life in severe lab or harsh plant environments. Hold button freezes the reading on the display. High-contrast 1 cm (3/8") LCD digits are easy to read. Minimum and maximum readings are available by pressing a button. Dual display shows humidity/temperature, humidity/dew point, barometric pressure/temperature, or barometric pressure/humidity.
Certificaciones: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Información para pedidos: Supplied with four AAA batteries.