VWR® Flat-Bottom Boiling Flasks, Short Neck
VW25055-500 VW25055-1000 VW25055-125 VW25055-300 VW25055-250
89000-328CS 6390.09 MXN
89000-336 89000-328 89000-330 89000-332 89000-334
VWR® Flat-Bottom Boiling Flasks, Short Neck
Borosilicate glass flasks have a full-length [ST] 24/40 joint and a short neck.
  • Borosilicate glass fabrication
Meet ASTM Specification E1403, Type I, Class 4 requirements.