Sterile Synthetic Gloves, Cardinal
Proveedor: Cardinal
Duraprene™
2Y72PN70 2Y72PN60 2Y72PN65 2Y72PN75 2Y72PN80 2Y72PN85 2Y72PN65 2Y72PN55 2Y72PN80 2Y72PN60 2Y72PN70 2Y72PN85 2Y72PN75 2Y72PN90 2Y72PN90 2Y72PN55
89027-166CS 13096.31 MXN
89027-162 CA89027-160 89027-164 CA89027-162 89027-170 89027-160 CA89027-170 CA89027-158 CA89027-168 89027-166 89027-156 CA89027-164 89027-168 CA89027-156 89027-158 CA89027-166
Sterile Synthetic Gloves, Cardinal
Guantes
These sterile cleanroom gloves provide a high level of comfort.
  • Nitrile and neoprene blend
  • Latex-free, powder-free
  • Hand specific, beaded cuff
Color: Dark yellow.