Falcon® Cell Scrapers, Sterile, Corning
Proveedor: Corning
Falcon®
353086 353085 353085 353086 353089 353087 353087 353089
15621-010CS 5800 MXN
15621-010 89008-786 CA15621-010 CA15621-015 CABD353089 CA15621-005 15621-015 15621-005
Falcon® Cell Scrapers, Sterile, Corning
Rascadores de células
PS handle, TPE blade. Designed to provide maximum accessibility to the growth surfaces of a variety of culture vessels.
  • Cross-ribbed PS handle provides greater rigidity, to ensure better control while scraping cells
  • Highly compliant TPE blade pivots to provide multiple angles to remove cells from the entire growth surface
Información para pedidos: Supplied individually wrapped in peel-open packages.