I-CHEM™ Boston Round Bottles, with Caps, Thermo Scientific
Proveedor: Thermo Scientific
229-0500 249-1000 149-1000 249-0250 329-0125 349-0500 229-0250 129-0125 229-0125 329-1000 349-1000 329-0500 229-1000 349-0250 149-0125 129-1000 249-0125 129-0250 149-0500 149-0250 329-0250 349-0125 249-0500 129-0500
IR329-0500CS 2260.98 MXN
IR129-0125 IR129-0500 IR329-0500 IR329-0125 IR229-0125 IR229-0500 IR349-0250 IR149-0500 IR149-0125 IR249-0250 IR249-1000 IR349-1000 IR149-1000 IR349-0500 IR149-0250 IR249-0125 IR349-0125 IR249-0500 IR229-1000 IR129-0250 IR329-0250 IR229-0250 IR329-1000 IR129-1000
I-CHEM™ Boston Round Bottles, with Caps, Thermo Scientific
Frascos Frascos para muestreo de agua
Amber and clear glass bottles for use in the analysis of semivolatiles, pesticides, PCB’s, and metals.
  • Convenient, sturdy bottles for general sampling and use
  • Amber glass bottles are designed for light-sensitive samples
  • I-CHEM™ 100 Series bottles, for general use, are not processed or certified
I-CHEM Certified™ 200 and 300 series bottles are processed to meet or exceed 'US EPA Specifications and Guidance for Contaminent-Free Sample Containers' and include sample-analysis labels, unattached. I-CHEM Certified™ 300 series bottles include a Certificate of Analysis with a bar-coded production number in every case.
Each bottle in the 300 series has a manually readable bar-code lot label for absolute traceability and for use with automated sample tracking systems.
Información para pedidos: Available in amber and clear glass. Supplied with fluoropolymer resin-lined polypropylene caps attached.