Round-Bottom Two-Neck Flasks, Angled Side Neck, Ace Glass
Proveedor: Ace Glass
6927-222 9463-206 9463-211 9463-210 9463-12 9463-212 9464-24 6927-204 9464-206 6927-208 6927-08 6927-226 9464-10 6927-66 6927-62 6927-20 9464-04 9464-295 6927-44 9464-210 9464-208 9464-06 6927-258 6927-68 6927-58 9463-06 9463-11 6927-262 6927-64 6927-239 9463-08 6927-04 6927-229 6927-160 6927-232 9463-10 9464-204 6927-220 6927-70 6927-32 6927-244 9464-08 6927-22 9463-208 6927-168
89057-152EA 2857.3 MXN
89074-762 89074-686 89074-760 89074-684 89074-766 89074-700 89057-200 89056-816 89074-764 89074-720 89074-704 89057-204 89074-724 89057-224 89074-708 89056-442 89056-464 89097-258 89056-460 89083-880 89056-440 89056-462 89056-826 89056-468 89056-842 89056-446 89057-152 89085-106 89074-734 89074-712 89074-710 89074-754 89097-260 89074-714 89083-874 89083-876 89083-878 89056-430 89056-454 89056-472 89056-438 89056-836 89056-434 89074-690 89056-436
Round-Bottom Two-Neck Flasks, Angled Side Neck, Ace Glass
Matraces Matraces de fondo redondo
Round-bottom flasks with angled side neck and [ST] outer joints
Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks. The outer joints are tooled with reinforcement rings for added strength and stability.