PYREX® VISTA™ Flasks, Flat Bottom, Glass, Clear, with Standard Tapered Joints, Corning
Proveedor: Corning
PYREX® VISTA™
70100-125 70100-500 70100-1L 70100-250
89090-350CS 10660 MXN
89090-350 89090-326 89090-388 89090-122
PYREX® VISTA™ Flasks, Flat Bottom, Glass, Clear, with Standard Tapered Joints, Corning
Matraces Matraces de fondo plano
Made of PYREX® borosilicate glass.
  • These flasks have full length outer 24/40 standard tapered joint
  • Wall thickness is controlled to provide balance between thermal expansion and mechanical strength for shock resistance
Reusable short neck boiling flask with flat bottom.
Certificaciones: ASTM E-1403