VWR® Disposable Anti-Static Polystyrene Pour Boats
Proveedor: VWR
VWR®
GPB-316
89106-760CS 8857.8 MXN
89106-760
VWR® Disposable Anti-Static Polystyrene Pour Boats
Navecillas Navecillas de pesaje
Strong, smooth polystyrene pour boats feature a single spout to facilitate dispensing.
  • Polystyrene construction
  • Antistatic properties
  • Stable and flat bottoms
Vessels are manufactured with an antistat treatment embedded into the plastic. This long-lasting treatment will not rub off or dissipate over time, making the boats suitable for use with static-affected materials. Stable and flat bottoms allow for easy stacking and convenient storage.