VWR® Countdown Controller
Proveedor: VWR
VWR®
5095
62379-567EA 1746.53 MXN
62379-567
VWR® Countdown Controller
Temporizadores Conmutadores de tiempo
Timer performs reproducible stirring, heating, or pumping times.
  • Traceable®
  • Countdown and Time-of-Day modes
  • Case is high-impact ABS plastic
Use for timed shutdown or start-up of apparatus during any test
A manual override function allows equipment to be switched on/off instantly without disturbing the program. Rechargeable NiCad battery maintains settings in the event of an electrical outage. Three-prong grounded receptacle relay controls equipment rated to 125 V, 15 A, 1800 W, ¹/₃ hp. A green LED indicates the on/off mode of attached equipment.
Programmable in any one-minute interval from 23 hours, 59 minutes to 1 minute. Countdown mode turns any piece of equipment on or off at the end of a set time period, which eliminates scheduling time-of-day settings and reduces programming time. Time-of-day mode permits programming of 14 a.m./p.m. clock settings for each day of the week, up to 196 on/off settings per week.
Certificaciones: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Información para pedidos: Supplied with rechargeable NiCad battery.