VWR® TraceClean® Cylinder Bottles, HDPE
Proveedor: VWR
VWR® TraceClean®
QLPY15900302 QLPY15900122 QLPY15900214 QLPY15900212 QLPY15900306 QLPY15900210 QLPY15900304 QLPY15900120 QLPY15900118
89094-076CS 858.86 MXN
89094-076 89094-088 89094-072 89094-084 89094-074 89094-090 89094-080 89094-092 89094-082
VWR® TraceClean® Cylinder Bottles, HDPE
Frascos Frascos para muestreo de agua
Plastic bottles are for use in metals analyses and multiple inorganic analytes.
  • HDPE construction
  • Suited to metals analyses
  • Supplied with polyfoam-lined caps
Processed (PC) and Quality-Assured® (QA) bottles follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA bottles include a Certificate of Analysis in every case, as well as bar codes with lot number and individual container number for complete traceability.
Not Processed (NPC) bottles do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.