VWR® TraceClean® Wide Mouth Jars, HDPE
Proveedor: VWR
VWR® TraceClean®
QLPS15900288 QLPS15900290 QLPS15900108 QLPS15900104 QLPS15900106 QLPS15900196 QLPS15900200 QLPS15900198 QLPS15900292
89094-098CS 981.33 MXN
89094-098 89094-110 89094-100 89094-094 89094-096 89094-106 89094-108 89094-102 89094-104
VWR® TraceClean® Wide Mouth Jars, HDPE
Frascos Frascos para muestreo de agua
Straight-sided jars are for collecting clays, slurries, and sediment matrices for use in geotechnical, environmental. Toxicity Characteristic Leaching Procedure (TCLP) metals, inorganic compounds, and benthic organism studies.
  • HDPE construction
  • Wide-mouthed design
  • Supplied with polyfoam-lined caps
Processed (PC) and Quality-Assured® (QA) jars follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA jars include a Certificate of Analysis in every case.
QA jars also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) jars do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.