VWR® TraceClean® Wide Mouth Packers, HDPE
Proveedor: VWR
VWR® TraceClean®
QLPW15900204 QLPW15900296 QLPW15900112 QLPW15900116 QLPW15900298 QLPW15900206 QLPW15900208 QLPW15900114 QLPW15900110 QLPW15900300
89094-112CS 943.73 MXN
89094-132 89094-122 89094-112 89094-134 89094-130 89094-118 89094-124 89094-114 89094-126 89094-116
VWR® TraceClean® Wide Mouth Packers, HDPE
Frascos Frascos para muestreo de agua
Packers are for use in collection of water samples for metals analyses and a variety of inorganic analytes including cyanide, sulfide, sulfate, chloride, and ion chromatography.
  • HDPE construction
  • Wide-mouthed design
  • Supplied with polyfoam-lined caps
Processed (PC) and Quality-Assured® (QA) packers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A "Specifications and Guidance for Contaminant-Free Sample Containers." QA packers include a Certificate of Analysis in every case.
QA packers also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) packers do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.