VWR® TraceClean® Wide Mouth Jars, Glass
Proveedor: VWR
VWR® TraceClean®
QLFM15900264 QLFM15900170 QLFM15900078 QLFM15900160 QLFM15900266 QLFM15900080 QLFM15900254 QLFM15900158 QLFS15900072 QLFM15900070 QLFS15900164 QLFM15900162 QLFS15900256 QLFM15900250 QLFM15900066 QLFM15900252 QLFM15900172 QLFM15900068
89094-014CS 2150.7 MXN
89093-992 89093-960 89094-008 89094-014 89094-004 89094-006 89094-010 89094-000 89094-012 89094-002 89093-958 89093-998 89093-944 89093-996 89093-952 89093-942 89093-950 89093-994
VWR® TraceClean® Wide Mouth Jars, Glass
Frascos Frascos para muestreo de agua
Type III (soda-lime) jars are for collection of solid matrices or liquids dispensed from bailers or peristaltic tubing.
  • Soda-lime glass fabrication
  • Wide-mouthed design
  • Available with solid-top or open-top caps
  • Individually numbered for traceability
  • Jars are available with solid-top or open-top caps
Processed (PC) and Quality-Assured® (QA) jars follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA jars include a Certificate of Analysis in every case.
QA jars also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) jars do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Solid-top caps with fluoropolymer resin liner are for use in semi-volatiles, pesticides, and PCBs; and select inorganic compounds. Open-top caps with ultrasonically bonded fluoropolymer resin/silicone septa are for use in volatile organic analysis.