VWR® TraceClean® Straight-Sided Wide Mouth Jars, Glass
Proveedor: VWR
VWR® TraceClean®
QLFS15900060 QLFS15900058 QLAS15900274 QLAS15900270 QLFS15900156 QLFS15900242 QLAS15900178 QLFS15900260 QLFS15900246 QLFS15900064 QLFS15900076 QLAS15900082 QLFS15900074 QLFS15900154 QLFS15900248 QLFS15900166 QLFS15900244 QLFS15900152 QLFS15900062 QLAS15900268 QLFS15900150 QLFS15900168 QLFS15900262 QLAS15900084
89093-970CS 2003.83 MXN
89093-970 89093-982 89093-990 89093-980 89094-018 89094-016 89094-028 89094-032 89094-024 89093-938 89094-030 89093-948 89093-978 89093-956 89093-968 89093-946 89093-954 89093-976 89093-988 89093-974 89093-986 89093-972 89093-984 89093-940
VWR® TraceClean® Straight-Sided Wide Mouth Jars, Glass
Frascos Frascos para muestreo de agua
Type III (soda-lime) glass jars are for collection of soil, sediment, and slurry matrices.
  • Amber glass fabrication
  • Straight sides
  • Wide-mouthed design
  • Recommended for UV light-sensitive samples
  • Jars are available with solid-top or open-top caps
Processed (PC) and Quality-Assured® (QA) jars follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA jars include a Certificate of Analysis in every case.
QA jars also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) jars do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.
Solid-top caps feature fluoropolymer resin liner and are for use in semi-volatiles, pesticides, and PCB analyses; and select inorganic compounds.
Open-top caps feature ultrasonically bonded fluoropolymer resin/silicone septum and are for use in head space volatile organic analysis. Amber glass jars are for use in semi-volatiles, pesticides, and PCB analyses; and select inorganic parameters for sediment and slurry matrices.