VWR® TraceClean® Jugs, Amber Glass
Proveedor: VWR
VWR® TraceClean®
QLAN15900100 QLAN15900102 QLAN15900286 QLAN15900192 QLAN15900284
89094-061CS 2186.28 MXN
89094-066 89094-061 89094-062 89094-058 89094-069
VWR® TraceClean® Jugs, Amber Glass
Frascos Frascos para muestreo de agua
Jugs are for use in semi-volatiles, pesticides, and PCBs analyses.
  • Amber glass is recommended for UV light-sensitive samples
  • Used in a variety of analyses
  • Completely traceable
  • Includes fluoropolymer resin-lined caps
Processed (PC) and Quality-Assured® (QA) jugs follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'.
QA jugs also include bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) jugs do not receive the specialty cleaning process.