Powder-Free Static-Dissipative Rolled Finger Cots, Honeywell Safety
Proveedor: Honeywell Safety Products
125LBR/M
56613-735CS 45762.9 MXN
56613-735
Powder-Free Static-Dissipative Rolled Finger Cots, Honeywell Safety
Protectores de dedos Protectores para dedos de látex
These multiple-washed finger cots are very low in particulates
Inherently static-dissipative, the powder-free finger cots will dissipate 5000 to 0 volts in 0.01 second (which exceeds the 2.0 seconds specified by MIL-B-817-5B). The average surface resistivity is 2x106ohms/sq.
Certificaciones: These long black finger cots meet the requirements of ASTM D3772 for physical properties, and the conductive bag packaging complies with MIL-P-82646A.