KIMBLE® RAY-SORB® Amber Class A Volumetric Flask, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMBLE® RAY-SORB®
28016-500 28016-250 28016-1000 28016-10 28016-200 28016-100 28016-50 28016-25
89002-014CS 9506.9 MXN
89002-022 89002-012 89002-014 89002-020 89002-010 89002-008 89002-016 89002-018
KIMBLE® RAY-SORB® Amber Class A Volumetric Flask, DWK Life Sciences
Matraces Matraces aforados
Calibrated to contain
Designed from ASTM Specification E288, Class A unserialized requirements.
RAY-SORB® process protects materials sensitive to light of the shorter wavelengths. Resistant to mechanical and thermal shock. Supplied with a [ST] PTFE stopper and color-coded handle. Flasks include a white marking spot.