Thermocouple Probes for Fluke® 51 and 52 Series II Thermometers, Fluke Corporation
Proveedor: Fluke
874610 750422 2098684 779942
16001-338EA 5247.65 MXN
16001-338 61221-105 61221-070 61221-160
Thermocouple Probes for Fluke® 51 and 52 Series II Thermometers, Fluke Corporation
Pruebas de temperatura
For use with Fluke 51 Series II (61221-038) and 52 Series II (61221-040) Thermometers.