NeoPro® Powder-Free Neoprene Gloves, Microflex®, Ansell
Proveedor: Ansell
NeoPro®
NPG-888-XS NPG-888-XL NPG-888-M NPG-888-XL NPG-888-S NPG-888-S NPG-888-M NPG-888-L NPG-888-L NPG-888-XS
32916-788CS 6780 MXN
32916-788 CA32916-796 CA32916-788 CA32916-792 32916-792 CA32916-794 32916-790 32916-796 CA32916-790 32916-794
NeoPro® Powder-Free Neoprene Gloves, Microflex®, Ansell
Guantes
Gloves are made entirely from a synthetic material with comfort close to latex that provides unique protection against a variety of chemicals.
  • Synthetic material with comfort close to latex, provides unique protection against variety of chemicals
  • Exceptional film consistency, manual dexterity, tactile sensitivity, reduced hand fatigue
  • Extended beaded cuff, ideal for high-risk environments
  • Textured fingertips for secure grip
  • Thickness: 6.7 mil (finger)
They offer exceptional film consistency, manual dexterity, tactile sensitivity, fine motor skill function, and reduced hand fatigue. Textured for a more secure grip.
Certificaciones: Gloves meet ASTM D 6319-99 standards.
Envase: Packaged 100 gloves per dispenser box.
Precaución: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.