Nalgene® Volumetric Flasks with Screw Cap, Polypropylene, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
4000-0100 4000-0050 4000-0200 4000-1000 4000-0500 4000-0250
29615-007CS 14764.23 MXN
29615-007 29615-018 29615-029 29615-062 29615-040 29615-030
Nalgene® Volumetric Flasks with Screw Cap, Polypropylene, Thermo Scientific
Matraces Matraces aforados
Flasks are lighter in weight than glass and break resistant to protect against leakage and contamination
Each flask is individually hand calibrated to contain and to deliver at 20°C (68°F). Flasks contain a permanent color graduation line and oval marking spot for ease in reading fluid levels. Autoclavable, leakproof, and graduated.
Certificaciones: Flasks meet ASTM E288 and ISO 384 Class B tolerances for consistent quality and dependability.