Providing dexterity with superb cut resistance for optimal productivity, gloves feature a polyurethane coating dipped on a liner structure of Dyneema® and LYCRA®.
- Palm coating for good grip
- Comfortable wear with great dexterity
- High level of abrasion and cut resistance
This innovative combination delivers a high level of abrasion resistance along with cut protection, while also preventing cuts and snags to the back of the hand.
Uses include aeronautics; assembling metal parts and components; assembly of white goods; finishing works; glass manufacturing; handling of sharp-edged objects, pre-assembling, cutting of small dry or lightly oiled parts and maintenance; and plastics injection and molding.
The palm coating provides dry grip and longer wear for confidence in handling small, sharp objects. Relaxed, natural-fit liner helps reduce hand fatigue. The polyurethane coating and liner combination improves tactile sensitivity, and has excellent grip in dry environments with effective control in minimally oily environments.
Certificaciones: ANSI Level 2 cut protection. ANSI Level 6 abrasion protection.
Envase: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.