VWR® Traceable® Infrared Thermometer with Wristband
Proveedor: VWR
VWR®
93677-74
36934-176EA 2929.9 MXN
36934-176
VWR® Traceable® Infrared Thermometer with Wristband
Termómetros Termómetros digitales
Non-invasive, no-touch measurements are ideal for food preparation, life sciences, pharmaceuticals, petroleum products, cleanrooms, electronics, and field use.
  • Traceable®
  • Laser beam pointer
  • Readings in both Celsius and Fahrenheit
  • Simple operation
  • Memory feature retains minimum/maximum readings
Simple operation allows unit to take readings in less than a second
Field of view is 6:1; thermometer can read a spot of 5 cm (2") from 30.5 cm (12") away. The emissivity of 0.05–1.00 in .01 steps covers most applications. Minimum/maximum feature allows viewing highest and lowest readings at any time. Laser beam permits perfect sample target sighting. Features a handy wristband. Display indicates low battery.
Unit takes both Celsius and Fahrenheit readings of solids, semi-solids, and liquids. Hold freezes the display at the current reading.
Certificaciones: A serial-numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate traceability to standards provided by the National Institute of Standards and Technology.
Información para pedidos: Supplied with button cell battery.