VWR® Type K Thermocouple Probes
Proveedor: VWR
VWR®
90028-29 90028-35
61161-372EA 1041.2 MXN
61161-372 61161-305
VWR® Type K Thermocouple Probes
Pruebas de temperatura
For use with all Type-K thermometers in liquids, semi-solids, and air/gas.
Note: Type K probes are not ISO/IEC 17025 accredited