KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, Glass Stopper, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMBLE® RAY-SORB®
92822G-500 92822G-10 92822G-100 92822G-200 92822G-25 92822G-50 92822G-2000 92822G-5 92822G-1000 92822G-20
89079-258CS 1556.99 MXN
89079-258 89079-246 89079-248 89079-250 89079-260 89079-252 89079-240 89079-242 89079-256 89079-244
KIMBLE® RAY-SORB® Amber Heavy Duty Wide Mouth Volumetric Flask, Glass Stopper, DWK Life Sciences
Matraces Matraces aforados
Flasks are made of red-tinted glass that provides protection from UV light
All flasks are graduated to Class A volumetric tolerances. Flasks with 5–250mL capacity are calibrated to meet ASTM Specification E288.
Feature wide mouth for easy pipet access and heavy, uniform walls for strength, durability, and safety. Interior bottoms are flat, allowing for efficient stirring with standard stir bars. Large, permanent markings allow for quick, accurate identification.