Nalgene® Sample Bags, Low-Density Polyethylene, Thermo Scientific
Proveedor: Thermo Scientific
Nalgene®
6255-0406 6255-0918 6255-0613 6255-0913 6255-0508
56766-016CS 10682.36 MXN
56766-130 56766-107 56766-071 56766-016 56766-049
Nalgene® Sample Bags, Low-Density Polyethylene, Thermo Scientific
Bolsas Bolsas para muestras
Transparent LDPE sample bags with strong, 4 mil walls are ideal for collecting, storing, sealing and protecting specimens, labware and small parts.
Sturdy 'zipper' locks securely and keeps dust out. Not recommended for storing liquids.