VWR® Extreme Temperature Mitt
Proveedor: Avantor
VWR®
HS1034AV
89026-680CS 18160.8 MXN
89026-680
VWR® Extreme Temperature Mitt
Protectores de manos
Full-hand, silicone rubber mitt is ideal for handling extremely hot or cold objects, from –57 to 260 °C (–70 to 500 °F), for brief periods of time.
  • Ideal for handling extremely hot or cold beakers, flasks, bottles, and similar items
  • Studded, oversized surface and internal ribs ensure safe grip
  • Silicone rubber mitt is designed to comfortably fit your hand
  • Withstands temperatures from –57 to 260 °C (–70 to 500 °F) for brief periods of time
Mitt features a studded, oversized surface and internal ribs to ensure a safe grip on beakers, flasks, bottles, and similar items.