KIMAX® Solution Bottles with Color-Coded PTFE [ST] Flathead Stopper, Kimble Chase, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMAX®
15097-100 15097-2000 15097-250 15097-500 15097-1000
89001-584CS 14517.24 MXN
89001-576 89001-584 89001-582 89001-580 89001-578
KIMAX® Solution Bottles with Color-Coded PTFE [ST] Flathead Stopper, Kimble Chase, DWK Life Sciences
Frascos Botellas con solucion
Narrow-mouth bottles conform to ASTM E438, Type 1, Class A standards.
Bottle necks are [ST] ground to accept flathead PTFE stoppers.