VWR® Wide Mouth Safety Wash Bottles, LDPE
Proveedor: VWR
VWR®
11646-0002 11646-2621 11646-2623 11646-2628 11646-2458 11646-3232 11646-2612 11646-2627 11646-2132 11646-2631 11646-2624 11646-2639 11646-2620 11646-2622 11646-2614 11646-2250 11646-2611 11646-2613
16649-950PK 841.07 MXN
16649-950 89141-046 16649-940 16649-918 16649-917 16649-925 16649-935 16649-905 16649-915 16649-943 16649-910 89141-042 16649-945 16649-901 16649-966 16649-955 16649-911 16649-900
VWR® Wide Mouth Safety Wash Bottles, LDPE
Frascos Frascos de lavado
Easy-to-read, break-resistant dispensing bottles include DOT, CAS numbers, National Fire Protection Association (NFPA) codes and symbols, protective equipment symbols, and target organ information.
  • Easy-to-read, low-density polyethylene dispensing bottles are break-resistant
  • Intended for short-term dispensing use
  • Include DOT, CAS numbers, NFPA codes and symbols, protective equipment symbols, and target organ information
Polypropylene nozzle assemblies are leakproof and color-coded to match bottle label.
Toluene bottle is solid red with white lettering and hazard information.Sodium Hypochlorite bottle is white LDPE for light-protection between 230 and 450 nm. Variety pack includes one each: acetone (red closure), ethanol (natural closure), isopropanol (yellow closure), methanol (green closure), toluene (red closure), and water (blue closure).
Caution: LDPE wash bottles are intended for short term dispensing use only and not for storage of acetone, dichloromethane, toluene, and 1,1,1-trichloroethane.
Certificaciones: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.