VWR® Safety-Vented™ Wide Mouth Labeled Wash Bottles, LDPE
Proveedor: VWR
VWR® Safety-Vented™
11642-2611 11642-2622 11642-2638 11643-6239 11642-2623 11642-2639 11642-2612 116422622 116422638 11642-2624
66001-147PK 729.78 MXN
89141-044 66001-147 66001-148 89141-048 66001-152 66001-141 66001-145 CA66001-155 CA66001-141 66001-155
VWR® Safety-Vented™ Wide Mouth Labeled Wash Bottles, LDPE
Frascos Frascos de lavado
Wide mouth, color-coded, low-density polyethylene wash bottles feature a unique two-piece polypropylene venting assembly that prevents pressure buildup of solvents inside the bottle.
  • Pre-labeled and vented for maximum safety
  • Eliminate solvent drips
  • Easy-to-read labeling
Venting assembly eliminates solvent drips from the spout due to internal evaporation. Easy-to-read solvent labeling includes DOT, NFPA codes and symbols, as well as CAS numbers. Bottles are leakproof.
Certificaciones: Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
Precaución: LDPE wash bottles are intended for short-term dispensing use only and not for long-term storage. Bottles marked LYOB (Label Your Own Bottle) are designed for use only with solvents compatible with LDPE.